Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900546

Sigma-Aldrich

Ethyl dodecanoate

Vetec, reagent grade, 98%

Synonym(s):

Ethyl laurate, Lauric acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOC2H5
CAS Number:
Molecular Weight:
228.37
Beilstein/REAXYS Number:
1769671
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

bp

269 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)OCC

InChI

1S/C14H28O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h3-13H2,1-2H3

InChI key

MMXKVMNBHPAILY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Botsi et al.
Carbohydrate research, 241, 37-46 (1993-03-17)
The inclusion complexes of cyclomaltoheptaose (beta CD) and heptakis(2,3,6-tri-O-methyl)cyclomaltoheptaose (TM-beta CD) with the four major components of the pheromone of the olive fruit fly (Dacus oleae), namely 1,7-dioxaspiro[5.5]undecane, (-)-alpha-pinene, nonanal, and ethyl dodecanoate, and the complex of heptakis(2,6-di-O-methyl)cyclomaltoheptaose (DM-beta CD)
Lianli Li et al.
International journal of pharmaceutics, 237(1-2), 77-85 (2002-04-17)
An ethyl laurate-based microemulsion system with Tween 80 as surfactant, propylene glycol and ethanol as cosolvents was developed for intranasal delivery of diazepam. Phase behavior and solubilization capacity of the microemulsion system were characterized and in vivo nasal absorption of
Hydrocracking of ethyl laurate on bifunctional micro-/mesoporous zeolite catalysts.
Oliver Busse et al.
ChemSusChem, 3(5), 563-565 (2010-05-04)
D R Bevan et al.
Cancer letters, 57(2), 173-179 (1991-05-01)
Effects of vehicles and of asbestos on disposition of benzo[a]pyrene (B[a]P) were examined in vivo. [3H]B[a]P dissolved in triethylene glycol, ethyl laurate, or tricaprylin was intratracheally administered to male Sprague-Dawley rats. Excretion was most rapid when triethylene glycol was the
Napaporn Komesvarakul et al.
Journal of cosmetic science, 57(4), 309-325 (2006-09-08)
Microemulsification of triglyceride-based oil is challenging due to the formation of undesirable phases such as macroemulsions, liquid crystals, or sponge phases. This research evaluates the formation of artificial sebum microemulsions using linker molecules, with the addition of co-oil to help

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service