Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900542

Sigma-Aldrich

Diethyl malonate

Vetec, reagent grade, 98%

Synonym(s):

Malonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOC2H5)2
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
774687
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.52 (vs air)

vapor pressure

1 mmHg ( 40 °C)

product line

Vetec

assay

98%

refractive index

n20/D 1.413 (lit.)

bp

199 °C (lit.)

mp

−51-−50 °C (lit.)

density

1.055 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CC(=O)OCC

InChI

1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3

InChI key

IYXGSMUGOJNHAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Marrone et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(43), 11537-11550 (2009-09-25)
The condensation of dialkyl beta-diesters with various aldehydes promoted by TiCl4 has been studied by DFT approaches and experimental methods, including NMR, IR and UV/Vis spectroscopy. Various possible reaction pathways have been investigated and their energy profiles evaluated to find
Chunying Shu et al.
Journal of the American Chemical Society, 129(50), 15710-15717 (2007-11-22)
The first reactions of trimetallic nitride templated endohedral metallofullerenes (TNT EMFs) with carbon radicals generated from diethyl malonate catalyzed by manganese(III) acetate are reported. Two methano monoadducts, Sc3N@C80-A and Sc3N@C80-B, were isolated and characterized. Sc3N@C80-A contains two ester moieties, whereas
Renhua Fan et al.
The Journal of organic chemistry, 72(15), 5905-5907 (2007-06-26)
A highly efficient, one-flask tandem Knoevenagel-Michael addition reaction of sulfonimines with diethyl malonate in the presence of a catalytic amount of base affords the corresponding arylidene dimalonates in good to excellent yields.
Hua Li et al.
Nucleosides, nucleotides & nucleic acids, 29(8), 581-590 (2010-07-28)
This article describes a very simple route for synthesizing novel lipophilic phosphonate bis(t-bu-SATE) prodrugs of acyclic cyclopentenylated nucleosides such as adenine 17 and cytosine 18. The key intermediate 6 was constructed via a ring-closing metathesis of compound 5, which could
Shigeyuki Yamada et al.
Organic & biomolecular chemistry, 5(9), 1442-1449 (2007-04-28)
The addition of heteroaromatic lithium reagents 2 to a THF solution of perfluorocyclopentene (1) provided preferentially the corresponding monosubstituted products 5, while the addition of 1 to 2 effectively gave the 1,2-disubstituted products 6 in good to excellent yields. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service