Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900540

Sigma-Aldrich

p-Tolualdehyde

Vetec, reagent grade, 97%

Synonym(s):

4-Methylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CHO
CAS Number:
Molecular Weight:
120.15
Beilstein/REAXYS Number:
385772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

97%

refractive index

n20/D 1.545 (lit.)

bp

204-205 °C (lit.)
82-85 °C/11 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(C)cc1

InChI

1S/C8H8O/c1-7-2-4-8(6-9)5-3-7/h2-6H,1H3

InChI key

FXLOVSHXALFLKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Ohkubo et al.
Chemical communications (Cambridge, England), 46(4), 601-603 (2010-01-12)
Photooxygenation of p-xylene by oxygen occurs efficiently under photoirradiation of 9-mesityl-2,7,10-trimethylacridinium ion (Me(2)Acr(+)-Mes) to yield p-tolualdehyde and hydrogen peroxide, which is initiated via photoinduced electron transfer of Me(2)Acr(+)-Mes to produce the electron-transfer state.
K Watanabe et al.
Drug metabolism and disposition: the biological fate of chemicals, 23(2), 261-265 (1995-02-01)
Mouse hepatic microsomal enzymes catalyzed the oxidation of o-, m-, and p-tolualdehydes, intermediate metabolites of xylene, to the corresponding toluic acids. Cofactor requirement for the catalytic activity indicates that the microsomes contain NAD- and NADPH-dependent enzymes for this reaction. GC/MS
Selvam Amudhan Senthan et al.
Dalton transactions (Cambridge, England : 2003), 44(33), 14813-14822 (2015-07-30)
A new series of octanuclear Ru(ii) complexes of tolylterpyridine appended calixresorcarenes [{Ru(ttpy)}8()](PF6)16 (), [{Ru(ttpy)}8()](PF6)16 (), and [{Ru(ttpy)}8()](PF6)16 () [ = 2,8,14,20-tetraethyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene; = 2,8,14,20-tetraphenyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene; and = 2,8,14,20-tetra-p-tolyl-4,6,10,12,16,18,22,24-octa(4'-p-benzyloxy-(2,2':6',2''-terpyridinyl))calix[4]resorcarene] have been synthesized and characterized. The tetraethyl-, tetraphenyl-, and tetra-p-tolylcalixresorcarenes (), (), and ()

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service