Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900538

Sigma-Aldrich

Cinnamyl alcohol

Vetec, reagent grade, 98%

Synonym(s):

3-Phenyl-2-propen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCH2OH
CAS Number:
Molecular Weight:
134.18
Beilstein/REAXYS Number:
1903999
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.6 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

product line

Vetec

assay

98%

bp

250 °C (lit.)

mp

30-33 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

OC\C=C\c1ccccc1

InChI

1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-7,10H,8H2/b7-4+

InChI key

OOCCDEMITAIZTP-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Nicolaou et al.
Journal of the American Chemical Society, 133(21), 8134-8137 (2011-05-06)
The development of an enantioselective allylic alcohol dichlorination catalyzed by dimeric cinchona alkaloid derivatives and employing aryl iododichlorides as chlorine sources is reported. Reaction optimization, exploration of the substrate scope, and a model for stereoinduction are presented.
C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(6), 837-866 (2005-04-07)
A toxicologic and dermatologic review of cinnamyl alcohol when used as a fragrance ingredient is presented.
H Ott et al.
Skin pharmacology and physiology, 23(4), 213-224 (2010-05-01)
Allergic contact dermatitis is a complex syndrome and knowledge about the in vitro detection of small-molecular-weight compounds, particularly prohaptens, is limited. Therefore, we investigated chemical-induced gene expression changes in human antigen-presenting cells upon stimulation with immunogenic contact allergens, prohaptens and
Carmen Martínez et al.
Journal of molecular graphics & modelling, 28(2), 196-201 (2009-08-04)
The free radical reactivity of monolignols can be explained in terms of the Fukui function and the local hard and soft acids and bases (HSAB) principle to determine the potential linkages among them for reactions involving free radicals. Our results
Annette Zeller et al.
Chemical research in toxicology, 22(12), 1929-1937 (2009-11-17)
The metabolism of the potent carcinogen estragole was investigated in humans after consumption of fennel tea by analyses of its metabolites in blood plasma and urine. Stable isotope dilution assays based on LC-MS/MS detection revealed that 1'-hydroxylation of estragole happened

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service