Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900536

Sigma-Aldrich

4-Methoxyphenylacetic acid

Vetec, reagent grade, 98%

Synonym(s):

Homoanisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein/REAXYS Number:
1101737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

140 °C/3 mmHg (lit.)

mp

84-86 °C (lit.)

SMILES string

COc1ccc(CC(O)=O)cc1

InChI

1S/C9H10O3/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

NRPFNQUDKRYCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

379.4 °F

flash_point_c

193 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Todoriki et al.
Journal of chromatography, 310(2), 273-281 (1984-10-12)
This paper describes the use of a high-performance liquid chromatograph equipped with an ultraviolet multi-detection system for the analysis of aromatic acids to help establish a high-risk screening system for disorders of organic acid metabolism. The peak height ratios of
Wai Mun Loke et al.
The Journal of nutrition, 139(12), 2309-2314 (2009-10-09)
Flavonoids are phytochemicals that are widespread in the human diet. Despite limitations in their bioavailability, experimental and epidemiological data suggest health benefits of flavonoid consumption. Valid biomarkers of flavonoid intake may be useful for estimating exposure in a range of
Wojciech Ostrowski et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 967, 21-27 (2014-07-27)
Liquid chromatography coupled to mass spectrometry (MS) with electrospray ionization (ESI) is one of analytical techniques to obtain accurate results of low molecular weight aromatic compounds in biological samples of different origin. The interpretations of mass spectra of these aromatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service