Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900526

Sigma-Aldrich

3-Pyridinemethanol

Vetec, reagent grade, 98%

Synonym(s):

ω-Hydroxy-3-picoline, 3-(Hydroxymethyl)pyridine, 3-Pyridyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein/REAXYS Number:
107851
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.545 (lit.)

bp

154 °C/28 mmHg (lit.)

density

1.124 g/mL at 25 °C (lit.)

SMILES string

OCc1cccnc1

InChI

1S/C6H7NO/c8-5-6-2-1-3-7-4-6/h1-4,8H,5H2

InChI key

MVQVNTPHUGQQHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Fazekas et al.
Acta physiologica Hungarica, 74(2), 169-174 (1989-01-01)
The antilipolytic, nicotinic acid analogue beta-pyridylcarbinol (Ronicol) has previously been reported to decrease the free fatty acid (FFA) concentration of the arteria-blood, and to moderate the FFA-uptake and O2-consumption of the myocardium; on this basis, the drug may be expected
T B Gelhard et al.
Journal de biologie buccale, 16(2), 95-99 (1988-06-01)
The F- uptake in vivo in sound human enamel from fluoridated toothpastes is described. Ten participants brushed for about 1 min twice a day, four enamel specimens in a dental appliance over three week periods. The enamel specimens were prepared
D J Wise et al.
Archives of biochemistry and biophysics, 344(1), 176-183 (1997-08-01)
Haemophilus parasuis malate dehydrogenase ((S)-malate:NAD+ oxidoreductase; EC 1.1.1.37) isolated from cell sonicates was purified 584-fold to electrophoretic homogeneity with a 19% recovery and a specific activity of 222 units/mg protein. SDS-polyacrylamide gel electrophoresis and molecular exclusion chromatography indicated the purified
M Went
Zeitschrift fur die gesamte innere Medizin und ihre Grenzgebiete, 42(11), 313-316 (1987-06-01)
In 30 patients with organic disturbances of the arterial blood supply of the lower extremities the caudal epidural combination injection (one per cent procaine and radecol comp.) in comparison to the caudal epidural sympathetic blocking one per cent procaine) was
C Keller et al.
Klinische Wochenschrift, 66(14), 647-650 (1988-07-15)
We report two cases of unusual hepatic lesions following years of monotherapy with pyridylcarbinol (timespan) in a daily dose of 1.35 to 1.5 g for familial hypercholesterolemia. Mild elevations of liver enzymes correlated to a large tumor-like mass in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service