Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900525

Sigma-Aldrich

Cyclohexanemethanol

Vetec, reagent grade, 98%

Synonym(s):

(Hydroxymethyl)cyclohexane, Cyclohexyl methanol, Hexahydrobenzyl alcohol, NSC 5288

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2OH
CAS Number:
Molecular Weight:
114.19
Beilstein/REAXYS Number:
773712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.465 (lit.)

bp

181 °C (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCCC1

InChI

1S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8H,1-6H2

InChI key

VSSAZBXXNIABDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasunori Okada et al.
Organic letters, 9(8), 1573-1576 (2007-03-16)
[reaction: see text] Ethyl 1-thio-2,3,4,6-tetrakis-O-triisopropylsilyl-beta-d-glucopyranoside, ethyl 6-O-benzyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside, and ethyl 6-O-pivaloyl-1-thio-2,3,4-tris-O-triisopropylsilyl-beta-d-glucopyranoside induced highly beta-selective O-glucosidations. Among them, the 6-O-pivaloylated substrate provided the best selectivity up to alpha/beta = 3:97 with cyclohexylmethanol, and the substrate was used for glucosidations with secondary and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service