Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900524

Sigma-Aldrich

Terephthalic acid

Vetec, reagent grade, 98%

Synonym(s):

Benzene-1,4-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein/REAXYS Number:
1909333
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

reagent grade

vapor pressure

<0.01 mmHg ( 20 °C)

product line

Vetec

assay

98%

form

powder

autoignition temp.

925 °F

mp

>300 °C (lit.)

solubility

water: ~0.017 g/L at 25 °C

density

1.58 g/cm3 at 25 °C

SMILES string

OC(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

KKEYFWRCBNTPAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingqi Tian et al.
Biosensors & bioelectronics, 71, 1-6 (2015-04-17)
Considerable recent attention has been paid to homogeneous fluorescent DNA detection with the use of nanostructures as a universal "quencher", but it still remains a great challenge to develop such nanosensor with the benefits of low cost, high speed, sensitivity
Na Chang et al.
Journal of chromatography. A, 1257, 116-124 (2012-08-28)
Metal-organic frameworks (MOFs) which offer a variety of topologies, porous networks and high surface areas are promising and have potential for the applications of specific adsorption, isomerization, catalysis and separation. UIO-66 is the first MOF that has been observed to
Osamu Kawakami et al.
Neurosurgery, 56(5), 1075-1081 (2005-04-28)
The objective of this study was to develop a therapeutic system of aneurysm embolization on the basis of organization of fibrous tissue. The healing process of aneurysms by means of polyethylene terephthalate (PET) fiber coils seeded with autologous fibroblasts was
Y H Kim et al.
Science (New York, N.Y.), 284(5414), 647-650 (1999-04-24)
Human recombinant tissue plasminogen activator (tPA) may benefit ischemic stroke patients by dissolving clots. However, independent of thrombolysis, tPA may also have deleterious effects on neurons by promoting excitotoxicity. Zinc neurotoxicity has been shown to be an additional key mechanism
Jasmine D Patel et al.
Journal of biomedical materials research. Part A, 73(3), 359-366 (2005-04-01)
Persistence of infection can occur when the host immune response is compromised because of the presence of a foreign implant. Surface modification of biomaterials with phospholipid polymers may enhance biocompatibility and reduce incidence of infection by impeding bacterial and leukocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service