Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900523

Sigma-Aldrich

p-Anisic acid

Vetec, reagent grade, 98%

Synonym(s):

4-Methoxybenzoic acid, p-Methoxybenzoic acid, Draconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
508910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

182-185 °C (lit.)

SMILES string

COc1ccc(cc1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ZEYHEAKUIGZSGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

365.0 °F - closed cup

flash_point_c

185 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gadgoli et al.
Journal of ethnopharmacology, 66(2), 187-192 (1999-08-05)
p-Methoxy benzoic acid isolated from the methanolic soluble fraction of the aqueous extract of Capparis spinosa L. (Capparidaceae) was found to possess significant antihepatotoxic activity against carbontetrachloride and paracetamol induced hepatotoxicity in vivo and thioacetamide and galactosamine induced hepatotoxicity in
N Chaves et al.
Journal of chemical ecology, 27(3), 611-621 (2001-07-10)
Eleven allelochemicals (ferulic acid, cinnamic acid, 4-hydroxybenzoic acid, hydroxycinnamic acid, methyl propionate, oxalic acid, methylmalonic acid, p-anisic acid, butyric acid, 3-hydroxybutyric acid, and azulene) were identified in the exudate of Cistus ladanifer L. We studied the effect of each on
T Ogiso et al.
Journal of pharmaceutical sciences, 87(5), 594-598 (1998-05-20)
The pharmacokinetics of aniracetam (AP), a new cognitive performance enhancer, and its main metabolites was investigated after intravenous (iv) and oral administrations to rat. The plasma levels of AP, 4-p-anisamidobutyric acid (ABA), and p-anisic acid (AA) were determined simultaneously by
Stephen G Bell et al.
Dalton transactions (Cambridge, England : 2003), 41(28), 8703-8714 (2012-06-15)
The crystal structures of the 4-methoxybenzoate bound forms of cytochrome P450 enzymes CYP199A2 and CYP199A4 from the Rhodopseudomonas palustris strains CGA009 and HaA2 have been solved. The structures of these two enzymes, which share 86% sequence identity, are very similar
H Twilfer et al.
European journal of biochemistry, 267(19), 5926-5934 (2000-09-21)
Using 4-methoxybenzoate monooxygenase from Pseudomonas putida, the substrate deuterium isotope effect on product formation and the solvent isotope effect on the stoichiometry of oxygen uptake, NADH oxidation, product and/or H2O2 (D2O2) formation for tight couplers, partial uncouplers, and uncouplers as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service