Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900517

Sigma-Aldrich

Nicotinamide

Vetec, reagent grade, ≥98%

Synonym(s):

Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
383619
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥98%

form

powder

color

white

mp

128-131 °C (lit.)

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

Inchi Key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitchell E Horwitz et al.
The Journal of clinical investigation, 124(7), 3121-3128 (2014-06-10)
Delayed hematopoietic recovery is a major drawback of umbilical cord blood (UCB) transplantation. Transplantation of ex vivo-expanded UCB shortens time to hematopoietic recovery, but long-term, robust engraftment by the expanded unit has yet to be demonstrated. We tested the hypothesis
Kathrin Schmeisser et al.
Nature chemical biology, 9(11), 693-700 (2013-10-01)
Sirtuins, a family of histone deacetylases, have a fiercely debated role in regulating lifespan. In contrast with recent observations, here we find that overexpression of sir-2.1, the ortholog of mammalian SirT1, does extend Caenorhabditis elegans lifespan. Sirtuins mandatorily convert NAD(+)
Thomas J Herzog et al.
Gynecologic oncology, 130(1), 25-30 (2013-04-18)
Sorafenib, an oral multikinase inhibitor of the VEGFR/PDGFR/Raf/MEK/ERK pathway, has shown potential activity in patients with recurrent ovarian cancer (OC). One strategy to prolong disease control and survival in patients with OC is maintenance therapy after achieving a complete response.
Robert J Motzer et al.
The Lancet. Oncology, 14(6), 552-562 (2013-04-20)
In a phase 3 trial comparing the efficacy and safety of axitinib versus sorafenib as second-line treatment for metastatic renal cell carcinoma, patients given axitinib had a longer progression-free survival (PFS). Here, we report overall survival and updated efficacy, quality
Salvatore Simmini et al.
Nature communications, 5, 5728-5728 (2014-12-17)
The endodermal lining of the adult gastro-intestinal tract harbours stem cells that are responsible for the day-to-day regeneration of the epithelium. Stem cells residing in the pyloric glands of the stomach and in the small intestinal crypts differ in their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service