Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900515

Sigma-Aldrich

Coumarin

Vetec, reagent grade, ≥99%

Synonym(s):

1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
383644
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

0.01 mmHg ( 47 °C)

product line

Vetec

assay

≥99%

bp

298 °C (lit.)

mp

68-73 °C (lit.)

SMILES string

O=C1Oc2ccccc2C=C1

InChI

1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H

InChI key

ZYGHJZDHTFUPRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

323.6 °F - closed cup

flash_point_c

162 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donglei Yu et al.
Medicinal research reviews, 23(3), 322-345 (2003-03-21)
Numerous plant-derived compounds have been evaluated for inhibitory effects against HIV replication, and some coumarins have been found to inhibit different stages in the HIV replication cycle. This review article describes recent progress in the discovery, structure modification, and structure-activity
Yutao Yang et al.
Biosensors & bioelectronics, 47, 300-306 (2013-04-17)
The synthesis and characterization of a coumarin-chromene (8, 9-dihydro-2H-cyclopenta[b]pyrano[2,3-f]chromene-2,10(7aH)-dione) (1) derivative and its use for thiol chemosensing in water was reported. Experimental details showed 1 acts as a probe for the detection of thiols including cysteine (Cys), homocysteine (Hcy) and
A ratiometric fluorescent probe for rapid detection of hydrogen sulfide in mitochondria.
Yuncong Chen et al.
Angewandte Chemie (International ed. in English), 52(6), 1688-1691 (2013-01-05)
Musiliyu A Musa et al.
Current medicinal chemistry, 15(26), 2664-2679 (2008-11-11)
The coumarin (benzopyran-2-one, or chromen-2-one) ring system, present in natural products (such as the anticoagulant warfarin) that display interesting pharmacological properties, has intrigued chemists and medicinal chemists for decades to explore the natural coumarins or synthetic analogs for their applicability
Sukhendu Maiti et al.
Journal of the American Chemical Society, 135(11), 4567-4572 (2013-03-07)
We present here, the design, synthesis, spectroscopic characterization, and in vitro biological assessment of a gemcitabine-coumarin-biotin conjugate (5). Probe 5 is a multifunctional molecule composed of a thiol-specific cleavable disulfide bond, a coumarin moiety as a fluorescent reporter, gemcitabine (GMC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service