Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900513

Sigma-Aldrich

Xylan

Vetec, reagent grade, 90%

Synonym(s):

Poly(β-D-xylopyranose[1→4])

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

90%

form

powder

mp

300 °C

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changjie Bao et al.
Scientific reports, 10(1), 11686-11686 (2020-07-17)
Saccharomyces cerevisiae-based expression systems, which rely on safe, food-grade strains, are low cost, simple to operate, and can be used for large-scale fermentation. However, low levels of foreign protein expression by S. cerevisiae have limited their widespread application. The ability
Masashi Kiyohara et al.
Journal of biochemistry, 140(3), 369-373 (2006-08-08)
Recently we reported the molecular cloning and characterization of a novel beta-1,3-xylanase from the marine bacterium Vibrio sp. AX-4 [Kiyohara et al. (2005) Biochem. J. 388, 949-957]. We report here the structural analysis of oligosaccharides generated from beta-1,3-xylan of a
Hiroshi Hashimoto et al.
FEBS letters, 579(20), 4324-4328 (2005-08-03)
Here, we present the crystal structure of the family 31 carbohydrate-binding module (CBM) of beta-1,3-xylanase from Alcaligenes sp. strain XY-234 (AlcCBM31) determined at a resolution of 1.25A. The AlcCBM31 shows affinity with only beta-1,3-xylan. The AlcCBM31 molecule makes a beta-sandwich
Shusen Wang et al.
Bioresource technology, 310, 123456-123456 (2020-05-11)
The aim of this work was to investigate the effects of cellulose, hemicellulose and lignin on the ignition behaviors of biomass. The ignition events of three components and five types of single biomass particles were captured by a high-speed camera
Masashi Kiyohara et al.
Journal of biochemistry, 146(5), 633-641 (2009-07-17)
beta-1,3-Xylanase from Vibrio sp. strain AX-4 (XYL4) is a modular enzyme composed of an N-terminal catalytic module belonging to glycoside hydrolase family 26 and two putative carbohydrate-binding modules (CBMs) belonging to family 31 in the C-terminal region. To investigate the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service