Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900494

Sigma-Aldrich

D-(+)-Glucosamine hydrochloride

Vetec, reagent grade, ≥99%

Synonym(s):

2-Amino-2-deoxy-D-glucose hydrochloride, Chitosamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Beilstein/REAXYS Number:
4157370
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

≥99%

mp

190-194 °C (dec.) (lit.)

SMILES string

Cl.N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C6H13NO5.ClH/c7-3-5(10)4(9)2(1-8)12-6(3)11;/h2-6,8-11H,1,7H2;1H/t2-,3-,4-,5-,6?;/m1./s1

InChI key

QKPLRMLTKYXDST-NSEZLWDYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glucosamine, an amino sugar, is the precursor of the hexosamine biosynthetic pathway leading to the formation of UDP-N-acetylglucosamine (UDP-GlcNAc), which is then used for making glycosaminoglycans, proteoglycans, and glycolipids. D-(+)-Glucosamine inhibits the coaggregation of Candida yeast species with the bacterial strain S. salivarius.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wan-Yu Chou et al.
Journal of cellular physiology, 230(9), 2240-2251 (2015-03-11)
Breast cancer is a common cancer leading to many deaths among females. Cyclooxygenase-2 (COX-2) and interleukin-8 (IL-8) are two highly expressed inflammatory mediators to be induced by the protein kinase C (PKC) signaling via various inflammatory stimuli and both contribute
Oshra Betzer et al.
ACS nano, 8(9), 9274-9285 (2014-08-19)
A critical problem in the development and implementation of stem cell-based therapy is the lack of reliable, noninvasive means to image and trace the cells post-transplantation and evaluate their biodistribution, final fate, and functionality. In this study, we developed a
Ji-Sun Hwang et al.
Metabolism: clinical and experimental, 64(3), 368-379 (2014-12-18)
This study investigated the potential of glucosamine (GlcN) to affect body weight gain and insulin sensitivity in mice normal and at risk for developing diabetes. Male C57BL/6J mice were fed either chow diet (CD) or a high fat diet (HFD)
Khadijeh Jamialahmadi et al.
Environmental toxicology and pharmacology, 38(1), 212-219 (2014-06-25)
Glucosamine (GlcN) is an important precursor in the biochemical synthesis of glycosylated proteins and lipids in human body. It gains importance because of its contribution to human health and its multiple biological and therapeutic effects. In this study, the in
Chunmei He et al.
Plant molecular biology, 88(3), 219-231 (2015-05-01)
Dendrobium officinale is a traditional Chinese medicinal plant. The stems of D. officinale contain mannan polysaccharides, which are promising bioactive polysaccharides for use as drugs. However, the genes involved in the biosynthesis of mannan polysaccharides in D. officinale have not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service