Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900491

Sigma-Aldrich

3-Indolepropionic acid

Vetec, reagent grade, 99%

Synonym(s):

3-(3-Indolyl)propanoic acid, IPA, Indole-3-propionic acid, NSC 3252, NSC 47831

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
Beilstein/REAXYS Number:
147733
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

134-135 °C (lit.)

SMILES string

OC(=O)CCc1c[nH]c2ccccc12

InChI

1S/C11H11NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2,(H,13,14)

InChI key

GOLXRNDWAUTYKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Fluorescent analogues of strigolactones
  • Anti-tumor agents
  • Melanocortin receptors ligands
  • Immunosuppressive agents
  • Iinhibitors of hepatitis C virus
  • Histamine H4 receptor agonists
  • NR2B/NMDA receptor antagonists
  • CB1 Antagonist for the treatment of obesity
  • Antibacterial agents
  • Inhibitor of TGF-β receptor binding

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youjia Yu et al.
Frontiers in physiology, 11, 65-65 (2020-03-03)
Paraquat (PQ) is a widely used herbicide which can cause high mortality to humans. However, relatively few studies focus on metabolic feature of PQ intoxication for investigating the underlying mechanisms. Here we performed non-targeted metabolomics profiling of serum samples from
In Koo Hwang et al.
Journal of neuroscience research, 87(9), 2126-2137 (2009-02-25)
Tryptophan-derived indole compounds have been widely investigated as antioxidants and as free-radical scavengers. Indole-3-propionic acid (IPA), one of these compounds, is a deamination product of tryptophan. In the present study, we used Mongolian gerbils to investigate IPA's neuroprotective effects against
K L Borden et al.
European journal of biochemistry, 202(2), 459-470 (1991-12-05)
The antirepressor indole 3-propanoate has been shown by X-ray crystallography to bind in a different orientation compared with the natural corepressor for the tryp repressor, L-tryptophan (Lawson, C.L. & Sigler, P. B. (1988) Nature 333, 869-871). This suggests a simple
Burkhard Poeggeler et al.
PloS one, 5(4), e10206-e10206 (2010-04-28)
Aging is a multi-factorial process, however, it is generally accepted that reactive oxygen species (ROS) are significant contributors. Mitochondria are important players in the aging process because they produce most of the cellular ROS. Despite the strength of the free-radical
R Hardeland et al.
Advances in experimental medicine and biology, 467, 389-395 (2000-03-18)
Chemiluminescence associated with oxidation by free radicals was investigated in an alkaline, hemin-catalysed hydrogen peroxide system, using the following tryptophan metabolites as radical scavengers: indole-3-pyruvic, indole-3-propionic, kynurenic, xanthurenic and quinaldic acids and 4-hydroxy-quinoline. Light emission from oxidation of the indolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service