Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900485

Sigma-Aldrich

L-α-Phosphatidylcholine

Vetec, reagent grade

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein/REAXYS Number:
5209585
EC Number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder or solid

functional group

phospholipid

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Guo et al.
Biochemical and biophysical research communications, 516(3), 914-920 (2019-07-06)
The Ig superfamily member V-domain Ig-containing suppressor of T-cell activation (VISTA) is a negative regulator with broad-spectrum activities and has reported that blockade of VISTA or combination with other negative checkpoint receptors sufficiently break tumor tolerance. However, it remains unclear
Cristian Dima et al.
Food chemistry, 195, 39-48 (2015-11-18)
The encapsulation by spray drying method of coriander essential oil (CEO) in various materials (chitosan, alginate, chitosan/alginate, chitosan/inulin) was studied. The viscoelastic properties of the oil-in-water (O/W) emulsions and the characteristics of CEO-loaded microcapsules like morphology, moisture, wettability, solubility, flowability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service