Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900475

Sigma-Aldrich

CAPSO

Vetec, reagent grade, ≥99%

Synonym(s):

3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid, CAPSO Free Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4S
CAS Number:
Molecular Weight:
237.32
Beilstein/REAXYS Number:
5939234
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥99%

form

crystalline powder

pH

2.5-4.0

useful pH range

8.9-10.3

pKa (25 °C)

9.6

solubility

water: 0.11 g/mL, clear, colorless

SMILES string

OC(CNC1CCCCC1)CS(O)(=O)=O

InChI

1S/C9H19NO4S/c11-9(7-15(12,13)14)6-10-8-4-2-1-3-5-8/h8-11H,1-7H2,(H,12,13,14)

InChI key

INEWUCPYEUEQTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Sun et al.
Antonie van Leeuwenhoek, 108(2), 291-299 (2015-06-13)
A Gram-stain negative, aerobic, non-motile and ovoid- to rod-shaped bacterial strain, designated JB3(T), was isolated from a wastewater sample collected from the biochemical reaction basin of Haiyan fine chemical factory in Zhejiang, China. Strain JB3(T) was found to grow optimally
D P McGregor et al.
BioTechniques, 21(3), 463-466 (1996-09-01)
The effect of buffer composition on simultaneous PCR amplification of 16S rRNA gene fragments of five bacterial species was examined using a number of different buffer systems. Tris-based PCR buffers at final concentrations of 10 mM proved unreliable. However, when
Paul M Mayer et al.
Physical chemistry chemical physics : PCCP, 13(11), 5178-5186 (2011-02-08)
The gas-phase binding of small molecules to the Amyloid β-40 peptide generated by electrospray ionization has been explored with collision-induced dissociation mass spectrometry and kinetic rate theory. This study discusses a simple procedure used to theoretically model the experimental breakdown
Etsuo A Susaki et al.
Nature communications, 11(1), 1982-1982 (2020-04-29)
Whole-organ/body three-dimensional (3D) staining and imaging have been enduring challenges in histology. By dissecting the complex physicochemical environment of the staining system, we developed a highly optimized 3D staining imaging pipeline based on CUBIC. Based on our precise characterization of
Mitsuhiro Okuda et al.
Biotechnology and bioengineering, 84(2), 187-194 (2003-09-11)
The iron storage protein, apoferritin, has a cavity in which iron is oxidized and stored as a hydrated oxide core. The size of the core is about 7 nm in diameter and is regulated by the cavity size. The cavity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service