Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900461

Sigma-Aldrich

MES sodium salt

Vetec, reagent grade, 99%

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
Beilstein/REAXYS Number:
3765682
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

crystalline powder

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

[Na+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Shirai et al.
Journal of molecular biology, 321(5), 879-889 (2002-09-11)
The crystal structure of congerin II, a galectin family lectin from conger eel, was determined at 1.45A resolution. The previously determined structure of its isoform, congerin I, had revealed a fold evolution via strand swap; however, the structure of congerin
Yue Yuan et al.
Biochemistry, 50(34), 7350-7360 (2011-08-03)
This study is aimed at investigating the molecular basis of environmental adaptation of woolly mammoth hemoglobin (Hb) to the harsh thermal conditions of the Pleistocene ice ages. To this end, we have carried out a comparative biochemical-biophysical characterization of the
Takamitsu Miyafusa et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 64(Pt 6), 512-515 (2008-06-10)
Capsular polysaccharides (CPs) are important virulence factors of Staphylococcus aureus. The biosynthesis of type 5 and type 8 CPs (CP5 and CP8), which are produced by most clinical isolates of S. aureus, is catalyzed by 16 CP-assembling proteins. One of
Theresa E Bankston et al.
Journal of chromatography. A, 1217(14), 2123-2131 (2010-03-03)
Hydroxyapatite (HAP) columns, widely used for chromatographic separation of proteins and other biomolecules because of their unique selectivity and ability to resolve complex mixtures, exhibit limited stability at acidic conditions requiring careful control of pH. Even with buffered solutions, however
Jin-Peng Sun et al.
The Journal of biological chemistry, 278(14), 12406-12414 (2003-01-28)
Protein-tyrosine phosphatase 1B (PTP1B) has been implicated as an important regulator in several signaling pathways including those initiated by insulin and leptin. Potent and specific PTP1B inhibitors could serve as useful tools in elucidating the physiological functions of PTP1B and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service