Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900450

Sigma-Aldrich

D-Glutamic acid

≥99%, powder, Vetec

Synonym(s):

(R)-2-Aminoglutaric acid, (R)-2-Aminopentanedioic acid, D-Glu, R-(−)-1-Aminopropane-1,3-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein/REAXYS Number:
1723800
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Product Name

D-Glutamic acid, Vetec, reagent grade, ≥99%

grade

reagent grade

product line

Vetec

assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

color

white to off-white

mp

200-202 °C (subl.) (lit.)

SMILES string

N[C@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m1/s1

InChI key

WHUUTDBJXJRKMK-GSVOUGTGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Unnatural isomer of glutamic acid.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Persson et al.
Molecular and cellular biology, 10(9), 4701-4711 (1990-09-01)
The gene encoding glutamic acid decarboxylase (GAD), the key enzyme in the synthesis of the inhibitory neurotransmitter gamma-aminobutyric acid, is shown to be expressed in the testis of several different species. Nucleotide sequence analysis of a cDNA clone isolated from
Jia Luo et al.
Science (New York, N.Y.), 298(5592), 425-429 (2002-10-12)
The motor abnormalities of Parkinson's disease (PD) are caused by alterations in basal ganglia network activity, including disinhibition of the subthalamic nucleus (STN), and excessive activity of the major output nuclei. Using adeno-associated viral vector-mediated somatic cell gene transfer, we
M I Mally et al.
Diabetologia, 39(4), 474-480 (1996-04-01)
Islet cell autoantigen 69 kDa (ICA69) has been reported as a polypeptide antigen expressed in pancreatic beta cells, and autoimmunity against this antigen has been associated with insulin-dependent diabetes mellitus. We have studied the cell type specificity and ontogeny of
Pierre Maechler et al.
Diabetes, 51 Suppl 1, S99-102 (2002-01-30)
It is intriguing that the kinetics of glucose-stimulated insulin secretion from the in situ perfused pancreas differ between the rat and the mouse. Here we confirm that insulin release in the rat is clearly biphasic, whereas in the mouse glucose
Katarzyna Rogoz et al.
Molecular pharmacology, 85(2), 322-334 (2013-11-28)
The transient receptor potential cation channel subfamily V member 1 (TRPV1) is known as a thermosensor and integrator of inflammation-induced hyperalgesia. TRPV1 is expressed in a subpopulation of primary afferent neurons that express several different neurotransmitters. The role of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service