Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900447

Sigma-Aldrich

TAPSO

Vetec, reagent grade, ≥99%

Synonym(s):

2-Hydroxy-3-[tris(hydroxymethyl)methylamino]-1-propanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-3-amino-2-hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO7S
CAS Number:
Molecular Weight:
259.28
Beilstein/REAXYS Number:
2113949
EC Number:
UNSPSC Code:
12161700
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥99%

form

powder

useful pH range

7.0-8.2

pKa (25 °C)

7.6

solubility

warm water: 0.33 g/mL, clear to slightly hazy, colorless

SMILES string

OCC(CO)(CO)NCC(O)CS(O)(=O)=O

InChI

1S/C7H17NO7S/c9-3-7(4-10,5-11)8-1-6(12)2-16(13,14)15/h6,8-12H,1-5H2,(H,13,14,15)

InChI key

RZQXOGQSPBYUKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

According to Ferreira, et al., TAPSO can have a strong interaction with iron in solution.[1] Based on this citation, it is recommended to use plastic-coated or PTFE-coated utensils when working with this chemical.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(Un)suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions - a review.
Ferreira C.M.H. et al.
Royal Society of Chemistry Advances, 5, 30989-31003 (2015)
(Un)suitability of the use of pH buffers in biological, biochemical and environmental studies and their interaction with metal ions ? a review.
Ferreira, C.M.H., et al.
Royal Society of Chemistry Advances, 5, 30989-31003 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service