Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900436

Sigma-Aldrich

D-Valine

Vetec, reagent grade, 98%

Synonym(s):

(R)-α-Aminoisovaleric acid, D-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
1721135
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

>295 °C (subl.) (lit.)

SMILES string

CC(C)[C@@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m1/s1

InChI key

KZSNJWFQEVHDMF-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly A Smith et al.
American journal of respiratory cell and molecular biology, 53(3), 355-367 (2015-01-09)
Hypoxic pulmonary vasoconstriction (HPV) is an important physiological response that optimizes the ventilation/perfusion ratio. Chronic hypoxia causes vascular remodeling, which is central to the pathogenesis of hypoxia-induced pulmonary hypertension (HPH). We have previously shown that Notch3 is up-regulated in HPH
Shanshan Song et al.
American journal of physiology. Cell physiology, 307(4), C373-C383 (2014-06-13)
An increase in cytosolic Ca(2+) concentration ([Ca(2+)]cyt) in pulmonary arterial smooth muscle cells (PASMC) is a major trigger for pulmonary vasoconstriction and an important stimulus for pulmonary arterial medial hypertrophy in patients with idiopathic pulmonary arterial hypertension (IPAH). Vascular smooth
Stefan M Gysler et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 73(3), 242-250 (2014-07-30)
Women with antiphospholipid syndrome (APS) are at increased risk of recurrent pregnancy loss (RPL) and preeclampsia. Antiphospholipid antibodies (aPL) directly alter trophoblast function. Treatment with low molecular weight heparin (LMWH) reduces the risk of RPL but not preeclampsia. Moreover, LMWH
Dean P J Kavanagh et al.
Stem cells (Dayton, Ohio), 33(9), 2785-2797 (2015-07-01)
Mesenchymal stem cells (MSCs) have shown therapeutic promise in many experimental and clinical models of inflammation. However, a commonly reported feature of MSC transplantation is poor homing to injured tissues. Previously, we have shown that pretreatment with cytokines/chemical factors enhances
Brian R Tuinema et al.
mBio, 5(6), e01886-e01886 (2014-11-27)
Neutrophils engulf and kill bacteria using oxidative and nonoxidative mechanisms. Despite robust antimicrobial activity, neutrophils are impaired in directing Salmonella clearance and harbor viable intracellular bacteria during early stages of infection that can subsequently escape to more-permissive cell types. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service