Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900426

Sigma-Aldrich

L-Tyrosine

≥98%, Vetec

Synonym(s):

(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
Beilstein/REAXYS Number:
392441
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

L-Tyrosine, Vetec, reagent grade, ≥98%

grade

reagent grade

product line

Vetec

assay

≥98%

form

powder

color

white to off-white

mp

>300 °C (dec.) (lit.)

SMILES string

N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

InChI key

OUYCCCASQSFEME-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Amino acid precursor of dopamine and other catecholamines

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hideki Hara et al.
Nature immunology, 14(12), 1247-1255 (2013-11-05)
The inflammasome adaptor ASC contributes to innate immunity through the activation of caspase-1. Here we found that signaling pathways dependent on the kinases Syk and Jnk were required for the activation of caspase-1 via the ASC-dependent inflammasomes NLRP3 and AIM2.
Keisuke Saito et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7690-7695 (2013-04-20)
Using quantum mechanics/molecular mechanics calculations and the 1.9-Å crystal structure of Photosystem II [Umena Y, Kawakami K, Shen J-R, Kamiya N (2011) Nature 473(7345):55-60], we investigated the H-bonding environment of the redox-active tyrosine D (TyrD) and obtained insights that help
Vincent Dunet et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(2), 207-214 (2012-02-04)
For the past decade, PET with (18)F-fluoro-ethyl-tyrosine ((18)F-FET) has been used in the evaluation of patients with primary brain tumors (PBTs), but so far series have reported only a limited number of patients. The purpose of this systematic review and
Verena Rahm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 546-550 (2014-02-26)
Low-grade gliomas (LGGs) may harbor malignant foci, which are characterized by increased tumor cellularity and angiogenesis. We used diffusion-weighted MR imaging (apparent diffusion coefficient [ADC]) and PET with the amino acid O-(2-(18)F-fluorethyl)-L-tyrosine ((18)F-FET) to search for focal changes of diffusion
Alena Volgusheva et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(18), 7223-7228 (2013-04-17)
Photobiological H2 production is an attractive option for renewable solar fuels. Sulfur-deprived cells of Chlamydomonas reinhardtii have been shown to produce hydrogen with the highest efficiency among photobiological systems. We have investigated the photosynthetic reactions during sulfur deprivation and H2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service