Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900423

Sigma-Aldrich

L-Histidine monohydrochloride monohydrate

Vetec, reagent grade, 99%

Synonym(s):

L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein/REAXYS Number:
4168261
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

254 °C (dec.) (lit.)

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Hendryk Gitter et al.
Pharmaceutics, 11(12) (2019-12-18)
In order to overcome the downside of long conventional freeze-drying (CFD) process times for monoclonal antibody formulations, microwave-assisted freeze-drying (MFD) was introduced. Recently, the general applicability and potential shortening of drying times were shown. However, little is known about the
Martin Maldonado et al.
Neurochemistry international, 85-86, 31-39 (2015-05-06)
The effect of reserpine on histamine (HA) and tele-methylhistamine (N(τ)-MHA) in hypothalamus and cortex of rats was analyzed and compared to catecholamines. IP injection of reserpine (5 mg/kg) confirmed the effectiveness of reserpine treatment on noradrenaline and dopamine levels. Our in-vitro
Verena Saller et al.
Journal of pharmaceutical sciences, 104(4), 1440-1450 (2015-01-22)
In a typical manufacturing setup for biopharmaceutical drug products, the fill and dosing pump is placed after the final sterile filtration unit in order to ensure adequate dispensing accuracy and avoid backpressure peaks. Given the sensitivity of protein molecules, peristaltic
Aldo Roda et al.
The Analyst, 139(24), 6494-6501 (2014-10-25)
Increasingly, smartphones are used as portable personal computers, revolutionizing communication styles and entire lifestyles. Using 3D-printing technology we have made a disposable minicartridge that can be easily prototyped to turn any kind of smartphone or tablet into a portable luminometer
Kristin Bjornsdottir-Butler et al.
Journal of food protection, 78(7), 1335-1342 (2015-07-22)
Recent developments in detection and enumeration of histamine-producing bacteria (HPB) have created powerful molecular-based tools to better understand the presence of spoilage bacteria and conditions, resulting in increased risk of scombrotoxin fish poisoning. We examined 235 scombrotoxin-forming fish from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service