Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900402

Sigma-Aldrich

Myristic acid

Vetec, reagent grade, 98%

Synonym(s):

1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
Beilstein/REAXYS Number:
508624
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

form

powder

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

>235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geok-Lin Chua et al.
PloS one, 7(7), e41924-e41924 (2012-07-31)
Integrins are signal transducer proteins involved in a number of vital physiological processes including cell adhesion, proliferation and migration. Integrin molecules are hetero-dimers composed of two distinct subunits, α and β. In humans, 18 α and 8 β subunits are
Alicja Kuban-Jankowska et al.
PloS one, 7(12), e52495-e52495 (2013-01-10)
Hydrogen peroxide induces oxidation and consequently inactivation of many protein tyrosine phosphatases. It was found that hydrogen peroxide, in the presence of carboxylic acids, was efficiently activated to form even more potent oxidant - peroxy acid. We have found that
Sheen X Lu et al.
Plant molecular biology, 82(3), 267-278 (2013-04-24)
Calcium-dependent protein kinases (CDPK) are a major group of calcium-stimulated kinases found in plants and some protists. Many CDPKs are membrane-associated, presumably because of lipid modifications at their amino termini. We investigated the subcellular location and myristoylation of AtCPK5, a
Anja Meier et al.
Hepatology (Baltimore, Md.), 58(1), 31-42 (2012-12-06)
Chronic infection with the human hepatitis B virus (HBV) is a global health problem and a main cause of progressive liver diseases. HBV exhibits a narrow host range, replicating primarily in hepatocytes. Both host and hepatocyte specificity presumably involve specific
S Curry et al.
Biochimica et biophysica acta, 1441(2-3), 131-140 (1999-11-26)
Human serum albumin possesses multiple fatty acid binding sites of varying affinities, but the precise locations of these sites have remained elusive. The determination of the crystal structure of human serum albumin complexed with myristic acid recently revealed the positions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service