Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900397

Sigma-Aldrich

Kinetin

Vetec, reagent grade, 99%

Synonym(s):

6-Furfurylaminopurine, N6-Furfuryladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N5O
CAS Number:
Molecular Weight:
215.21
Beilstein/REAXYS Number:
21703
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

264-270 °C (dec.) (lit.)

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccco3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Iwanaga et al.
Parasitology international, 62(5), 423-430 (2013-05-22)
The cell cycle of Plasmodium is unique among major eukaryotic cell cycle models. Cyclin-dependent kinases (CDKs) are thought to be the key molecular switches that regulate cell cycle progression in the parasite. However, little information is available about Plasmodium CDKs.
Mehdi Rajabi et al.
Nucleosides, nucleotides & nucleic acids, 31(6), 474-481 (2012-06-01)
Cytokinins and cytokinin nucleosides are purine derivatives with potential anticancer activity both in vitro and in vivo. N(6)-furfuryladenosine (kinetin riboside, KR) displays antiproliferative and apoptogenic activity against various human cancer cell lines and has recently been shown to suppress tumor
Natasha Spadafora et al.
Annals of botany, 110(8), 1631-1639 (2012-10-16)
How plant cell-cycle genes interface with development is unclear. Preliminary evidence from our laboratory suggested that over-expression of the cell cycle checkpoint gene, WEE1, repressed growth and development. Here the hypothesis is tested that the level of WEE1 has a
Jan Barciszewski et al.
International journal of biological macromolecules, 40(3), 182-192 (2006-08-11)
Cytokinins are important adenine derivatives that serve as hormones to control many processes in plants. They were discovered as factors that promote cell division in tobacco tissue cultures and have been shown also to regulate several other developmental events. Kinetin
Firdaus-e-Bareen et al.
Journal of hazardous materials, 237-238, 186-193 (2012-09-11)
"Assisted phytoextraction" involving application of chemical additives such as plant growth regulators (PGRs) has become a trend in phytoremediation technology. This study identifies a cost-effective, naturally available crude PGR (PGR1) that produces the same effects as the commercial PGR (PGR2)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service