Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900387

Sigma-Aldrich

2-Thiobarbituric acid

Vetec, reagent grade, ≥98%

Synonym(s):

4,6-Dihydroxy-2-mercaptopyrimidine, 4,6-Dihydroxypyrimidine-2-thiol, TBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2S
CAS Number:
Molecular Weight:
144.15
Beilstein/REAXYS Number:
120663
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥98%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

245 °C (dec.) (lit.)

SMILES string

O=C1CC(=O)NC(=S)N1

InChI

1S/C4H4N2O2S/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

Inchi Key

RVBUGGBMJDPOST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ismael Galván et al.
Physiological and biochemical zoology : PBZ, 85(4), 332-347 (2012-06-19)
The synthesis of melanins, which are the most common animal pigments, is influenced by glutathione (GSH), a key intracellular antioxidant. At high GSH levels, pheomelanin (the lightest melanin form) is produced, whereas production of eumelanin (the darkest melanin form) does
Sechang Oh et al.
Hepatology (Baltimore, Md.), 61(4), 1205-1215 (2014-10-02)
Recently, the beneficial effects of increased physical activity (PA) on nonalcoholic fatty liver disease (NAFLD) in obese subjects were reported. However, the optimal strength and volume of PA in lifestyle modification to improve NAFLD pathophysiology and be recommended as an
Fernando F Anhê et al.
Gut, 64(6), 872-883 (2014-08-01)
The increasing prevalence of obesity and type 2 diabetes (T2D) demonstrates the failure of conventional treatments to curb these diseases. The gut microbiota has been put forward as a key player in the pathophysiology of diet-induced T2D. Importantly, cranberry (Vaccinium
A P Moloney et al.
Meat science, 92(1), 1-7 (2012-04-21)
Colour and lipid stability of M. longissimus dorsi (LD) from sheep fed diets containing different lipid sources (Megalac (MG), camelina oil (CO), linseed oil (LO), NaOH-treated camelina seed (CS), NaOH-treated linseed (LS) or CO treated with ethanolamine (CA)) were examined.
Leticia R Benetti et al.
European journal of pharmacology, 698(1-3), 463-469 (2012-11-28)
Recent studies show that endogenous hydrogen sulfide (H(2)S) plays an anti-inflammatory role in the pathogenesis of airway inflammation. This study investigated whether exogenous H(2)S may counteract oxidative stress-mediated lung damage in allergic mice. Female BALB/c mice previously sensitized with ovalbumin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service