Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900384

Sigma-Aldrich

Indoxyl β-D-glucoside

Vetec, reagent grade, 97%

Synonym(s):

3-(β-D-Glucosido)indole, Plant indican

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO6
CAS Number:
Molecular Weight:
295.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

form

powder

solubility

ethanol: 50 mg/mL, clear to very slightly hazy

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccccc23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H17NO6/c16-6-10-11(17)12(18)13(19)14(21-10)20-9-5-15-8-4-2-1-3-7(8)9/h1-5,10-19H,6H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

XVARCVCWNFACQC-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thierry Maugard et al.
Biotechnology progress, 18(5), 1104-1108 (2002-10-05)
In this article, a HPLC method to identify and quantify the dyes and the indigo precursors produced in Polygonum tinctorium is described. Using this technique, indican has been positively identified in extracts of P. tinctorium. Our work with two cultivars
[Determination of indican in Polygonum tinctorium ait. in various growth periods].
H J Wang et al.
Zhong yao tong bao (Beijing, China : 1981), 13(11), 11-13 (1988-11-01)
R Armon et al.
Canadian journal of microbiology, 34(1), 78-79 (1988-01-01)
Medium m-CP, designed for the isolation of Clostridium perfringens from water samples, contains indoxyl beta-D-glucoside, an expensive chemical that is present at a high concentration in this medium. The use of m-CP with three concentrations of indoxyl beta-D-glucoside was tested
A N Ley et al.
Canadian journal of microbiology, 34(5), 690-693 (1988-05-01)
About 97% of Escherichia coli strains produce beta-glucuronidase, but almost all other Enterobacteriaceae lack this enzyme. A D-glucopyranosiduronic acid (glucuronide) possessing a readily detectable beta-linked aglycone should, therefore, constitute a specific reagent for the detection of this organism. For this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service