Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900383

Supelco

Linolenic acid

Vetec, reagent grade, 98%

Synonym(s):

α-Lnn, cis,cis,cis-9,12,15-Octadecatrienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7CO2H
CAS Number:
Molecular Weight:
278.43
Beilstein/REAXYS Number:
1727693
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

liquid

refractive index

n20/D 1.480 (lit.)

bp

230-232 °C/1 mmHg (lit.)

mp

-11 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

functional group

carboxylic acid
oleic acid

shipped in

ambient

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\CCCCCCCC(O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-

InChI key

DTOSIQBPPRVQHS-PDBXOOCHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An ω-3 fatty acid that serves as a precursor to eicosapentaenoic acid (EPA) but not docosahexaenoic acid. Conversion is greater in women than men, and conversely, β-oxidation metabolism is greater in men than women.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eoin Barrett et al.
PloS one, 7(11), e48159-e48159 (2012-11-28)
The aim of this study was to compare the impact of dietary supplementation with a Bifidobacterium breve strain together with linoleic acid & α-linolenic acid, for 7 weeks, on colonic sensitivity and fatty acid metabolism in rats. Maternally separated and
Gwendolyn Barceló-Coblijn et al.
Progress in lipid research, 48(6), 355-374 (2009-07-22)
There is little doubt regarding the essential nature of alpha-linolenic acid (ALA), yet the capacity of dietary ALA to maintain adequate tissue levels of long chain n-3 fatty acids remains quite controversial. This simple point remains highly debated despite evidence
Ju-Sheng Zheng et al.
BMJ (Clinical research ed.), 346, f3706-f3706 (2013-07-03)
To investigate the association between intake of fish and n-3 polyunsaturated fatty acids (n-3 PUFA) and the risk of breast cancer and to evaluate the potential dose-response relation. Meta-analysis and systematic review of prospective cohort studies. PubMed and Embase up
An Pan et al.
The American journal of clinical nutrition, 96(6), 1262-1273 (2012-10-19)
Prior studies of α-linolenic acid (ALA), a plant-derived omega-3 (n-3) fatty acid, and cardiovascular disease (CVD) risk have generated inconsistent results. We conducted a meta-analysis to summarize the evidence regarding the relation of ALA and CVD risk. We searched multiple
Joel A Simon et al.
The American journal of clinical nutrition, 89(5), 1558S-1564S (2009-03-27)
alpha-Linolenic acid (ALA; 18:3n-3) has been associated inconsistently with an increased risk of prostate cancer. Additional studies have become available since the publication of 2 previous meta-analyses. The objective was to review the published data on the relation between ALA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service