Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900371

Sigma-Aldrich

Spermine tetrahydrochloride

Vetec, reagent grade

Synonym(s):

N,N′-Bis(3-aminopropyl)-1,4-butanediamine tetrahydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H26N4 · 4HCl
CAS Number:
Molecular Weight:
348.18
Beilstein/REAXYS Number:
3911771
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

mp

310-311 °C (dec.) (lit.)

SMILES string

Cl.Cl.Cl.Cl.NCCCNCCCCNCCCN

InChI

1S/C10H26N4.4ClH/c11-5-3-9-13-7-1-2-8-14-10-4-6-12;;;;/h13-14H,1-12H2;4*1H

InChI key

XLDKUDAXZWHPFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to precipitate DNA from low salt aqueous buffers.

Biochem/physiol Actions

Mixed NMDA agonist/antagonist at the polyamine site. Neuroprotective effects have been observed at high concentrations (1 mM), while neurotoxicity is observed at lower concentrations. It enhances agonist effectiveness at the strychnine-insensitive glycine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA agonist/antagonist at the polyamine site. Plays a role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).
Mixed NMDA agonist/antagonist at the polyamine site. Role in cellular proliferation and differentiation; inhibits neuronal nitric oxide synthase (nNOS).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Desiree Bailey et al.
Pharmacology, biochemistry, and behavior, 133, 57-64 (2015-04-01)
The aim of this study was to examine the acute effect of a range of novel hydroxycinnamic acid derivatives of spermine on the development of spermine-induced CNS excitation and convulsions in female Laca mice, and to assess the chronic adverse
Enzo Agostinelli et al.
International journal of oncology, 45(3), 1109-1122 (2014-06-28)
It has been confirmed that multidrug resistant (MDR) melanoma cells (M14 ADR2) are more sensitive than their wild-type counterparts (M14 WT) to H2O2 and aldehydes, the products of bovine serum amine oxidase (BSAO)-catalyzed oxidation of spermine. The metabolites formed by
Alexis Goichon et al.
The American journal of clinical nutrition, 102(2), 359-367 (2015-06-26)
Amino acids are well known to be key effectors of gut protein turnover. We recently reported that enteral delivery of proteins markedly stimulated global duodenal protein synthesis in carbohydrate-fed healthy humans, but specifically affected proteins remain unknown. We aimed to
Tsuyoshi Minami et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(7), 721-724 (2015-07-15)
This study is the first to report on the detection of biogenic amines in an aqueous solution using an organic field-effect transistor (OFET) device with an extended gate electrode modified with a layer of diamine oxidase and a horseradish peroxidase
Julia Torres et al.
Organic & biomolecular chemistry, 13(27), 7500-7512 (2015-06-13)
Within all the eukaryotic cells myo-inositol phosphates (InsPs) are an important group of biomolecules that are potentially related to signaling functions. The most abundant member of this family in nature is InsP6 (phytate, L(12-) in its fully deprotonated form). The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service