V900368
Calcium D-gluconate
Vetec™, reagent grade, 98%
Synonym(s):
2,3,4,5,6-Pentahydroxycaproic Acid hemicalcium salt, Gluconic acid hemicalcium salt
About This Item
grade
reagent grade
product line
Vetec™
assay
98%
form
powder
SMILES string
OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Ca]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
InChI
1S/2C6H12O7.Ca/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;/h2*2-5,7-11H,1H2,(H,12,13);/q;;+2/p-2/t2*2-,3-,4+,5-;/m11./s1
InChI key
NEEHYRZPVYRGPP-IYEMJOQQSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service