Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900353

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine monohydrochloride

Vetec, reagent grade, 95%

Synonym(s):

4-(Dimethylamino)aniline monohydrochloride, 4-Amino-N,N-dimethylaniline monohydrochloride, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2·HCl
CAS Number:
Molecular Weight:
172.66
Beilstein/REAXYS Number:
4546912
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

95%

mp

215 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.ClH/c1-10(2)8-5-3-7(9)4-6-8;/h3-6H,9H2,1-2H3;1H

InChI key

KTWNIUBGGFBRKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Resp. Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kraiwan Punyain et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 368-378 (2011-09-20)
Organic radical cations form dicationic dimers in solution, observed experimentally as diamagnetic species in temperature-dependent EPR and low temperature UV/Vis spectroscopy. Dimerization of paraphenylenediamine, N,N-dimethyl-paraphenylenediamine and 2,3,5,6-tetramethyl-paraphenylenediamine radical cation in ethanol/diethylether mixture was investigated theoretically according to geometry, energetics and
Joseph B Issa et al.
The journal of physical chemistry. B, 111(24), 6878-6886 (2007-06-02)
Photoinduced intramolecular charge separation across proline-bridged donor-acceptor complexes of the type Pyr-(Pro)n-DMPD (where Pyr=pyrene-1-sulfonyl and DMPD=N,N-dimethyl-1,4-phenylenediamine) was studied. The steady-state emission spectrum for n=0, 1, 2, 3 showed an increase in emission intensity with the number of proline residues. Time-dependent
V Fogliano et al.
Journal of agricultural and food chemistry, 47(3), 1035-1040 (1999-12-20)
A novel method for measuring the antioxidant activity using N, N-dimethyl-p-phenylenediamine (DMPD) was developed. The radical cation of this compound gives a stable colored solution and a linear inhibition of color formation can be observed in the presence of 0.
F Tessier et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 66(2), 166-170 (1996-01-01)
An HPLC micro-method with fluorescence detection has been developed to determine total vitamin C (vit C) and dehydroascorbic acid (DHA) concentrations in human plasma samples. This method is based on the rapid, specific reaction of DHA with dimethyl-o-phenylenediamine (DMPD) to
K Wimalasena et al.
The Journal of biological chemistry, 271(42), 26032-26043 (1996-10-18)
The interactions of reductants with dopamine beta-monooxygenase (DbetaM) were examined using two novel classes of reductants. The steady-state kinetics of the previously characterized DbetaM reductant, N,N-dimethyl-1,4-p-phenylenediamine (DMPD), were parallel to the ascorbic acid-supported reaction with respect to pH dependence and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service