Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900344

Sigma-Aldrich

Ibuprofen

Vetec, reagent grade, 97%

Synonym(s):

α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

SMILES string

CC(C)Cc1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

Inchi Key

HEFNNWSXXWATRW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R O Day et al.
Clinical pharmacology and therapeutics, 43(5), 480-487 (1988-05-01)
The simultaneous disposition of the enantiomers of ibuprofen in synovial fluid and plasma was studied in eight patients with arthritis. Concentrations of the active S-enantiomer in synovial fluid exceeded those of the R-enantiomer at all times in all patients with
Bart Van Overmeire et al.
Lancet (London, England), 364(9449), 1945-1949 (2004-11-30)
Ibuprofen is used for treatment and prevention of patent ductus arteriosus in low-birthweight infants. Its effects on regional circulations differ from those of indometacin. Because prophylactic indometacin reduces the frequency of severe intraventricular haemorrhage and patent ductus arteriosus, we aimed
J B Whitlam et al.
Clinical pharmacology and therapeutics, 29(4), 487-492 (1981-04-01)
Free and total ibuprofen levels in serum and synovial fluid (SF) were determined in one male and 14 female arthritic patients (mean age 56 yr, range 19 to 77) after 400 mg three times daily for 2 days. Free drug
Carl Llor et al.
BMJ (Clinical research ed.), 347, f5762-f5762 (2013-10-08)
To evaluate the efficacy of oral anti-inflammatory or antibiotic treatment compared with placebo in the resolution of cough in patients with uncomplicated acute bronchitis and discoloured sputum. Multicentre, parallel, single blinded placebo controlled, randomised clinical trial. Nine primary care centres
Roland Neumann et al.
Neonatology, 102(1), 9-15 (2012-03-15)
Pharmacological closure of patent ductus arteriosus (PDA) is commonly achieved by intravenous (IV) administration of ibuprofen or indomethacin. Occasionally, oral ibuprofen is used for PDA treatment although its efficacy and safety are unclear. To systematically review randomized and quasi-randomized trials

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service