Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900332

Sigma-Aldrich

Chitin from shrimp shells

Vetec, reagent grade

Synonym(s):

Poly-(1→4)-β-N-acetyl-D-glucosamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

biological source

shrimp shells

grade

reagent grade

product line

Vetec

form

powder

SMILES string

CC(=O)N[C@@H]1[C@H](O)O[C@@H](CO)[C@H](O)[C@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4?,5?,6?,7?,8-/m1/s1

Inchi Key

OVRNDRQMDRJTHS-WTZNIHQSSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mitsumasa Osada et al.
Carbohydrate polymers, 92(2), 1573-1578 (2013-02-13)
This study examined the effects of a combined pretreatment with supercritical water and mechanochemical grinding with a ball mill on the physicochemical properties of chitin and its enzymatic degradation. Following pretreatment with a combination of supercritical water and grinding, chitin
Gustav Vaaje-Kolstad et al.
The FEBS journal, 280(13), 3028-3049 (2013-02-13)
The chitinolytic machinery of Serratia marcescens is one of the best known enzyme systems for the conversion of insoluble polysaccharides. This machinery includes four chitin-active enzymes: ChiC, an endo-acting non-processive chitinase; ChiA and ChiB, two processive chitinases moving along chitin chains
Zengxin Ma et al.
Carbohydrate polymers, 94(1), 272-277 (2013-04-03)
The effects of chitosan and oligachitosan on resistance induction of peach fruit against brown rot caused by Monilinia fructicola were investigated. Both chitosan and oligochitosan showed significant effect on controlling this disease. Moreover, chitosan and oligochitosan delayed fruit softening and
Dongli Guan et al.
Biotechnology and bioengineering, 110(9), 2471-2481 (2013-04-10)
Rapid and efficient tag removal remains a significant problem in recombinant protein purification. Using an engineered DnaE intein from Nostoc punctiforme, we developed a split intein mediated ultra-rapid purification (SIRP) method for the purification of tagless recombinant protein from E.
Flavia Laffleur et al.
Future medicinal chemistry, 5(5), 511-522 (2013-04-12)
The objective of this study was to develop and evaluate a more effective mucoadhesive thiomer for buccal drug-delivery systems. 2-iminothiolane was covalently attached to a chitosan backbone. A preactivation step followed, mediated by 6,6´dithionicotinamide, thiol groups were modified by disulfide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service