Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900323

Sigma-Aldrich

Hydroxyurea

Vetec, reagent grade, ≥98%

Synonym(s):

Hydroxycarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONHOH
CAS Number:
Molecular Weight:
76.05
Beilstein/REAXYS Number:
1741548
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥98%

storage temp.

2-8°C

SMILES string

NC(=O)NO

InChI

1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)

InChI key

VSNHCAURESNICA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anti-neoplastic. Inactivates ribonucleoside reductase by forming a free radical nitroxide that binds a tyrosyl free radical in the active site of the enzyme. This blocks the synthesis of deoxynucleotides, which inhibits DNA synthesis and induces synchronization or cell death in S-phase.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepa Manwani et al.
Blood, 122(24), 3892-3898 (2013-09-21)
Recurrent and unpredictable episodes of vaso-occlusion are the hallmark of sickle cell disease. Symptomatic management and prevention of these events using the fetal hemoglobin-reactivating agent hydroxyurea are currently the mainstay of treatment. Discoveries over the past 2 decades have highlighted
Winfred C Wang et al.
Pediatrics, 132(4), 677-683 (2013-09-04)
In the BABY HUG trial, young children with sickle cell anemia randomized to receive hydroxyurea had fewer episodes of pain, hospitalization, and transfusions. With anticipated broader use of hydroxyurea in this population, we sought to estimate medical costs of care
Guido Finazzi et al.
British journal of haematology, 161(5), 688-694 (2013-04-12)
Givinostat, a histone-deacetylase inhibitor (HDACi), inhibits proliferation of cells bearing the JAK2 V617F mutation and has shown significant activity with good tolerability in patients with chronic myeloproliferative neoplasms (MPN). In this multicentre, open-label, phase II study, 44 patients with polycythaemia
John J Strouse et al.
Pediatric blood & cancer, 59(2), 365-371 (2012-04-21)
Hydroxyurea is the only approved medication in the United States for the treatment of sickle cell anemia (HbSS) and is widely used in children despite an indication limited to adults. We review the evidence of efficacy and safety in children
Russell E Ware
Blood, 115(26), 5300-5311 (2010-03-13)
Hydroxyurea has many characteristics of an ideal drug for sickle cell anemia (SCA) and provides therapeutic benefit through multiple mechanisms of action. Over the past 25 years, substantial experience has accumulated regarding its safety and efficacy for patients with SCA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service