Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900321

Sigma-Aldrich

Adipic acid

Vetec, reagent grade, ≥99%

Synonym(s):

Hexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
1209788
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 159.5 °C)

product line

Vetec

assay

≥99%

autoignition temp.

788 °F

bp

265 °C/100 mmHg (lit.)

mp

151-154 °C (lit.)

SMILES string

OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

384.8 °F - closed cup

flash_point_c

196 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roohan Rakhshaee et al.
Journal of hazardous materials, 189(1-2), 158-166 (2011-03-15)
The removal of methylene blue (MB) as a cationic dye from aqueous solution by the stabilized Fe(3)O(4) nano-particles with the extracted pectin from apple waste (FN-PA) increased due to using the cross-linked forms of the bound pectin on the nano-particles
H V Pawar et al.
Colloids and surfaces. B, Biointerfaces, 102, 102-110 (2012-09-26)
Streptomycin (STP) and diclofenac (DLF) loaded film dressings were prepared by blending Polyox(®) (POL) with four hydrophilic polymers [hydroxypropylmethylcellulose (HPMC), carrageenan (CAR), sodium alginate (SA) or chitosan (CS)] using glycerol (GLY) as plasticiser. The films were characterised by scanning electron
Holger Noack et al.
Inorganic chemistry, 50(4), 1194-1202 (2011-01-28)
Adipic acid is a key compound in the chemical industry, where it is mainly used in the production of polymers. The conventional process of its generation requires vast amounts of energy and, moreover, produces environmentally deleterious substances. Thus, there is
Krzysztof J Paluch et al.
Journal of pharmaceutical sciences, 100(8), 3268-3283 (2011-04-08)
The production of salt or cocrystalline forms is a common approach to alter the physicochemical properties of pharmaceutical compounds. The goal of this work was to evaluate the impact of anion choice (succinate, adipate, and sulfate) on the physicochemical characteristics
Tânia Veiga et al.
Metabolic engineering, 14(4), 437-448 (2012-04-25)
Industrial production of semi-synthetic cephalosporins by Penicillium chrysogenum requires supplementation of the growth media with the side-chain precursor adipic acid. In glucose-limited chemostat cultures of P. chrysogenum, up to 88% of the consumed adipic acid was not recovered in cephalosporin-related

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service