Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900317

Sigma-Aldrich

Benzyldimethylhexadecylammonium chloride

Vetec, reagent grade, 97%

Synonym(s):

16-BAC, Benzylhexadecyldimethylammonium chloride, Cetalkonium chloride, Cetyldimethylbenzylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
396.09
Beilstein/REAXYS Number:
3802341
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mol wt

396.09 g/mol

mp

55-65 °C (lit.)

SMILES string

[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C25H46N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-26(2,3)24-25-21-18-17-19-22-25;/h17-19,21-22H,4-16,20,23-24H2,1-3H3;1H/q+1;/p-1

Inchi Key

SXPWTBGAZSPLHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

143.6 °F - Pensky-Martens closed cup

flash_point_c

62 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Morciano et al.
Journal of neurochemistry, 95(6), 1732-1745 (2005-11-05)
The nerve terminal proteome governs neurotransmitter release as well as the structural and functional dynamics of the presynaptic compartment. In order to further define specific presynaptic subproteomes we used subcellular fractionation and a monoclonal antibody against the synaptic vesicle protein
J Hartinger et al.
Analytical biochemistry, 240(1), 126-133 (1996-08-15)
Integral membrane proteins, particularly those with more than one transmembrane domain, have traditionally been difficult to separate by two-dimensional electrophoretic methods. Here we report the adaptation of a previously published procedure [D. E. Macfarlane (1989) Anal. Biochem. 176, 457-463] for
Prabhat K Singh et al.
Chemical communications (Cambridge, England), 47(24), 6912-6914 (2011-05-20)
The reactivity between two charged molecules and the activity of charged biomolecules are mainly governed by the principle of electrostatic interaction, i.e., like charges repel and opposite charges attract. In the present study it is shown that the principle of
H Liang et al.
Molecular vision, 14, 204-216 (2008-03-19)
To evaluate and compare the toxicological profiles of two quaternary ammonium compounds (QAC), benzalkonium chloride (BAK), and cetalkonium chloride (CKC), in standard solution or cationic emulsion formulations in rabbit eyes using newly developed in vivo and ex vivo experimental approaches.
Tom M H Choy et al.
Electrophoresis, 24(17), 3040-3048 (2003-09-16)
New insights into the effects of the wall adsorption of the visualization agent on baseline noise characteristics in indirect UV detection in capillary electrophoresis were provided. When compared to relatively small, hydrophilic cationic (1-(4-pyridyl)pyridinium chloride hydrochloride) or anionic chromophores (bromide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service