Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900314

Sigma-Aldrich

2-Naphthoxyacetic acid

Vetec, reagent grade, 98%

Synonym(s):

BNOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH2CO2H
CAS Number:
Molecular Weight:
202.21
Beilstein/REAXYS Number:
1074148
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

151-154 °C (lit.)

SMILES string

OC(=O)COc1ccc2ccccc2c1

InChI

1S/C12H10O3/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2,(H,13,14)

InChI key

RZCJYMOBWVJQGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Donbrow et al.
The Journal of pharmacy and pharmacology, 34(4), 215-224 (1982-04-01)
Stability constants and thermodynamic parameters have been evaluated for the complexation reaction in aqueous solution of caffeine with beta-naphthoxy acetic acid. The values were higher than those previously reported for the complexation of other ligands with methyl xanthines. In nearly
Tsai-Hui Duh et al.
Journal of chromatography. A, 987(1-2), 205-209 (2003-03-05)
In continuing the search for fluorescent reagents for analytical derivatization in chromatography, we found a simple chemical, (2-naphthoxy)acetyl chloride, with potential fluorophore/chromophore characteristics for the highly sensitive detection of analytes with an amino function. The reagent has an auxochrome (a
Chun Zhou et al.
Environmental monitoring and assessment, 154(1-4), 233-239 (2008-06-17)
A reliable and sensitive competitive fluorescence immunoassay for the quantitative determination of naphthalene (NA) was developed. 2-naphthoxy acetic acid (NAA) was selected as the hapten of naphthalene. Active ester method (AEM) was used to couple the NAA to carrier proteins
Hu Zhao et al.
Planta, 216(2), 293-301 (2002-11-26)
The plant hormone auxin affects cell elongation in both roots and shoots. In roots, the predominant action of auxin is to inhibit cell elongation while in shoots auxin, at normal physiological levels, stimulates elongation. The question of whether the primary
K A Rashid et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 21(3), 243-250 (1986-06-01)
Beta-naphthoxyacetic acid (BNOA) is used as a plant growth regulator on tomatoes and strawberries. It is the active ingredient in Blossom-Set and Berry-Set, two plant hormone sprays for fruit-set. The mutagenic activity of BNOA was evaluated in four strains of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service