Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900313

Sigma-Aldrich

Methyl salicylate

Vetec, reagent grade, 99%

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
971516
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

product line

Vetec

assay

99%

form

liquid

autoignition temp.

847 °F

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

204.8 °F - closed cup

flash_point_c

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard Radnovich et al.
Journal of pain research, 7, 727-735 (2014-12-20)
Treatment for pain due to shoulder impingement syndrome (SIS) typically begins conservatively with nonsteroidal anti-inflammatory drugs and physical therapy and can include subacromial injection of corticosteroids, particularly in patients unresponsive to conservative measures. The heated lidocaine/tetracaine (HLT) patch has been
Agata Bartyzel et al.
Analytica chimica acta, 707(1-2), 204-209 (2011-10-27)
N,N'-bis-(α-methylsalicylidene)-2,2-dimethyl-1,3-propanediimine (SBTD) modified silica gel was prepared and used as sorbent for solid phase extraction of Cr(III) ions from aqueous solution. This sorbent showed a high sorption affinity for Cr(III) while recovery of Cr(VI) was very low. The analyte ion
Michiel van Wijk et al.
PloS one, 6(7), e21742-e21742 (2011-07-19)
Insect herbivory induces plant odors that attract herbivores' natural enemies. Assuming this attraction emerges from individual compounds, genetic control over odor emission of crops may provide a rationale for manipulating the distribution of predators used for pest control. However, studies
Nadir Erbilgin et al.
Tree physiology, 32(8), 946-957 (2012-06-05)
Coniferous trees have both constitutive and inducible defences that deter or kill herbivores and pathogens. We investigated constitutive and induced monoterpene responses of jack pine (Pinus banksiana Lamb.) to a number of damage types: a fungal associate of the mountain
Youli Yao et al.
The Plant cell, 23(10), 3842-3852 (2011-10-27)
We have previously shown that local exposure of plants to stress results in a systemic increase in genome instability. Here, we show that UV-C-irradiated plants produce a volatile signal that triggers an increase in genome instability in neighboring nonirradiated Arabidopsis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service