Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900305

Sigma-Aldrich

Myristyltrimethylammonium bromide

Vetec, reagent grade, ≥99%

Synonym(s):

Tetradecyltrimethylammonium bromide, Trimethyl(tetradecyl)ammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13N(Br)(CH3)3
CAS Number:
Molecular Weight:
336.39
Beilstein/REAXYS Number:
3633227
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

≥99%

mol wt

micellar avg mol wt 27,000

aggregation number

80

CMC

4-5 mM (20-25°C)

mp

245-250 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C17H38N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18(2,3)4;/h5-17H2,1-4H3;1H/q+1;/p-1

InChI key

CXRFDZFCGOPDTD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

target_organs

Gastrointestinal tract, Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd Sajid Ali et al.
Journal of colloid and interface science, 352(2), 436-443 (2010-09-25)
Unfolding of rabbit serum albumin (RSA) by cationic surfactants cetyltrimethylammonium bromide (CTAB) and tetradecyltrimethylammonium bromide (TTAB) was studied by exploiting surface tensiometry, small-angle neutron scattering (SANS), intrinsic fluorescence, resonance Rayleigh scattering (RRS) (also referred as turbidity at 350/350), and circular
Suruchi Mahajan et al.
Langmuir : the ACS journal of surfaces and colloids, 28(50), 17238-17246 (2012-12-12)
Keeping in view the use of surfactants in drug delivery, the interactions of surface active ionic liquids, such as 1-tetradecyl-3-methylimidazolium bromide (C(14)mimBr), with drugs, viz., dopamine hydrochloride (DH) and acetylcholine chloride (AC), have been studied, and the results are further
Juan Zhang et al.
The journal of physical chemistry. B, 114(41), 13128-13135 (2010-09-28)
Two fatty acids, perfluorononanoic acid (C(8)F(17)COOH) and nonanoic acid (C(8)H(17)COOH), were mixed with a cationic hydrocarbon surfactant, tetradecyltrimethylammonium hydroxide (TTAOH), in aqueous solutions for comparative investigation. Phase behaviors of the two systems are quite different because of the special properties
Cexiong Fu et al.
Journal of chromatography. A, 1216(10), 1901-1907 (2009-02-03)
The usefulness of the micellar selectivity triangle (MST) for prediction and interpretation of separation patterns in micellar electrokinetic chromatography (MEKC) separations is presented. In addition, we demonstrate the capability of controlling selectivity properties of micelles through addition of organic modifiers
Changcheng Liu et al.
The journal of physical chemistry. B, 114(30), 9795-9804 (2010-07-14)
The phase behavior, rheological properties, and structures of two salt-free catanionic surfactant systems, tetradecyltrimethylammonium hydroxide ((TTA)OH)/lauric acid (LA)/H(2)O and cetyltrimethylammonium hydroxide ((CTA)OH)/LA/H(2)O, in the presence of deoxycholic acid (DeCA) were investigated and compared with the results of cholic acid (CA).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service