Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900299

Sigma-Aldrich

Trizma® phosphate dibasic

Vetec, reagent grade, 99%

Synonym(s):

Di(tris[hydroxymethyl]aminomethane) phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H11NO3)2 · H3PO4
CAS Number:
Molecular Weight:
340.27
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

99%

form

crystalline powder

solubility

water: 0.5 g/mL, clear, colorless

SMILES string

OP(O)(O)=O.NC(CO)(CO)CO.NC(CO)(CO)CO

InChI

1S/2C4H11NO3.H3O4P/c2*5-4(1-6,2-7)3-8;1-5(2,3)4/h2*6-8H,1-3,5H2;(H3,1,2,3,4)

InChI key

MYASHAYBKIUKNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service