V900284
Magnesium carbonate hydroxide hydrate
Vetec™, reagent grade, 99%
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C4Mg4O12 · H2MgO2 · xH2O
Molecular Weight:
395.58 (anhydrous basis)
EC Number:
UNSPSC Code:
12352300
grade
reagent grade
product line
Vetec™
assay
99%
form
powder
SMILES string
O.[Mg++].[Mg++].[Mg++].[Mg++].O[Mg]O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service