Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900280

Sigma-Aldrich

Tin(II) chloride dihydrate

Vetec, reagent grade, 98%

Synonym(s):

Stannous chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2 · 2H2O
CAS Number:
Molecular Weight:
225.65
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
powder, crystals or chunks

grade

reagent grade

product line

Vetec

assay

98%

form

powder, crystals or chunks

bp

652 °C (lit.)

mp

37-38 °C (dec.) (lit.)

Storage temp.

2-8°C

SMILES string

O.O.Cl[SnH2]Cl

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

Inchi Key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

target_organs

Cardio-vascular system, Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Hallett et al.
Organic & biomolecular chemistry, 10(30), 6130-6158 (2012-04-20)
Reactions of the allyltin trichloride 45 generated from (4S)-4-benzyloxypent-2-enyl(tributyl)stannane 1 with imines prepared from glyoxylates proceed with useful levels of 1,5-stereocontrol in favour of (4E)-2,6-anti-2-(alkylamino)-6-benzyloxyhept-4-enoates 49. This stereoselectivity, controlled by the chirality of the stannane, dominates over any intrinsic stereochemical
Grégory Dupeyre et al.
Organic & biomolecular chemistry, 9(22), 7780-7790 (2011-10-07)
A six-step one-pot reaction was designed for synthesizing homodimeric 7-phenylindolo[3,2-a]carbazoles from 1H-indoles and β-nitrostyrenes, in the presence of SnCl(2)·2H(2)O. The reactions proceeded under very mild conditions and the desired heterocycles were obtained in moderate to good yields. An unprecedented mechanism
Amit Pratap Singh et al.
Journal of the American Chemical Society, 134(10), 4998-5003 (2012-02-15)
Cationic and anionic species of heavier low-valent group 14 elements are intriguing targets in main group chemistry due to their synthetic potential and industrial applications. In the present study, we describe the synthesis of cationic (MCl(+)) and anionic (MCl(3)(-)) species
Liyan Zheng et al.
Dalton transactions (Cambridge, England : 2003), 41(5), 1630-1634 (2011-12-08)
It was found that stannous chloride (SnCl(2)), as a popular inorganic reducing reagent, could obviously enhance the electrochemiluminescence (ECL) of tris(2,2'-bipyridyl) ruthenium(II) (Ru(bpy)(3)(2+)) in aqueous solution. Some factors affecting the ECL reactions between Ru(bpy)(3)(2+) and Sn(2+), including pH, concentrations of
Aili Sun et al.
Talanta, 88, 259-264 (2012-01-24)
The Zn-Sn nanoparticles/multiwall carbon nanotubes (Zn-SnNPs/MWNTs) nanocomposite film was prepared by electrodeposition of ZnCl(2) and SnCl(2) on MWNTs simultaneously in Ethaline ionic liquids. Then, based on immobilizing hemoglobin (Hb) within a novel Zn-SnNPs/MWNTs nanocomposite film (Hb/Zn-SnNPs/MWNTs), a novel hydrogen peroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service