Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900278

Sigma-Aldrich

Iron(III) chloride hexahydrate

Vetec, reagent grade, 98%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
98%
grade:
reagent grade
form:
lumps
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

1 mmHg ( 194 °C)

product line

Vetec

assay

98%

form

lumps

bp

280-285 °C (lit.)

mp

37 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Kashyap et al.
ACS nano, 8(9), 9097-9106 (2014-08-28)
Biomineralization proteins are widely used as templating agents in biomimetic synthesis of a variety of organic-inorganic nanostructures. However, the role of the protein in controlling the nucleation and growth of biomimetic particles is not well understood, because the mechanism of
Dongying Ma et al.
Blood, 122(25), 4094-4106 (2013-10-26)
The identity of vampire bat saliva anticoagulant remained elusive for almost a century. Sequencing the salivary gland genes from the vampire bat Desmodus rotundus identified Desmolaris as a novel 21.5-kDa naturally deleted (Kunitz 1-domainless) form of tissue factor pathway inhibitor.
Xiaoqing Li et al.
The Journal of organic chemistry, 78(18), 9499-9504 (2013-08-28)
Halosulfonylation of terminal alkynes was achieved with sulfonylhydrazides as the sulfonyl precursor and inexpensive iron halide as halide source in the presence of TBHP, allowing the regio- and stereoselective generation of (E)-β-chloro and bromo vinylsulfones.
Ko-Lin Kuo et al.
PloS one, 7(12), e50295-e50295 (2012-12-12)
The association between intravenous (IV) iron administration and outcomes in hemodialysis (HD) patients is still debated. Therefore, this study was aimed to assess the relationship between the IV administration of ferric chloride hexahydrate (Atofen®) and cardiovascular (CV) outcome and the
Jinmyung Cha et al.
Chemical communications (Cambridge, England), 49(5), 457-459 (2012-10-27)
50 nm Magnetic nanocluster (MNC) was deliberately selected as an efficient probe for the magnetic relaxation switching (MRSw) assay. After surface modification of MNCs with poly(acrylic acid) to give PAA-MNCs, and employing streptavidin-biotin interactions for the MRSw assay model, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service