Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900277

Sigma-Aldrich

Lanthanum(III) chloride heptahydrate

Vetec, reagent grade, 65%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LaCl3 · 7H2O
CAS Number:
Molecular Weight:
371.37
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
reagent grade
form:
powder or crystals

grade

reagent grade

product line

Vetec

form

powder or crystals

concentration

65%

pH

~5 (25 °C, 100 g/L)

mp

91 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.O.O.Cl[La](Cl)Cl

InChI

1S/3ClH.La.7H2O/h3*1H;;7*1H2/q;;;+3;;;;;;;/p-3

Inchi Key

FDFPDGIMPRFRJP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Calcium channel blocker

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Citta et al.
Journal of inorganic biochemistry, 117, 18-24 (2012-10-20)
Lanthanum chloride (LaCl(3)) is a good inhibitor of both the cytosolic and mitochondrial thioredoxin reductase with IC(50) values of 1.75 and 7.46 μM, respectively. On the contrary, the related enzyme glutathione reductase is not inhibited by lanthanum ions even at
Yasuhiko Suzuki et al.
Biochemical and biophysical research communications, 410(2), 317-321 (2011-06-15)
Bradykinin (BK), a mediator of pain and inflammation, is involved in bone metabolism. We have previously reported that BK increased the synthesis of interleukin-6 and prostaglandin E(2) via phosphorylation of ERK1/2 in human osteoblasts, SaM-1. In the present study, we
Mario Pink et al.
Electrophoresis, 34(4), 501-504 (2012-11-30)
The recent introduction of the La(3+) precipitation method for the enrichment of phosphoproteins allows a gel-based analysis of these posttranslationally modified proteins. However, if this method is applied to cell lysates stored in urea-containing lysis buffer for an extended period
Paola Ciceri et al.
Biochemical and biophysical research communications, 418(4), 770-773 (2012-02-09)
Phosphate (Pi)-binders are commonly used in dialysis patients to control high Pi levels, that associated with vascular calcification (VC). The aim of this study was to investigate the effects of lanthanum chloride (LaCl(3)) on the progression of high Pi-induced VC
Mario Pink et al.
Journal of proteomics, 75(2), 375-383 (2011-08-30)
Posttranslational modification (PTM) of proteins, particularly phosphorylation, is a key element in the regulation of cell functions. In many signal transduction processes, PTM is a pivotal step. Various analytical methods have been proposed for the identification of phosphoproteins; however, most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service