Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900272

Sigma-Aldrich

Hydrazine sulfate salt

Vetec, reagent grade, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2SO4
CAS Number:
Molecular Weight:
130.12
Beilstein/REAXYS Number:
8778859
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
97%
grade:
reagent grade
form:
powder or crystals
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

form

powder or crystals

pH

1.3 (52 g/L)

mp

254 °C (lit.)

SMILES string

NN.OS(O)(=O)=O

InChI

1S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)

InChI key

ZGCHATBSUIJLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranendu Sekhar Das et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 4068-4080 (2013-01-24)
PVP capped platinum nano particles (PNP) of 5 nm diameter were prepared and characterized as homogeneous and of spherical nature. At physiological pH range (6.0-8.0), these PNP catalyze the deoxygenation of phenoxazine group containing resazurin (1) by hydrazine. The observed
Boran Kartal et al.
FEMS microbiology reviews, 37(3), 428-461 (2012-12-06)
Anaerobic ammonium-oxidizing (anammox) bacteria primarily grow by the oxidation of ammonium coupled to nitrite reduction, using CO2 as the sole carbon source. Although they were neglected for a long time, anammox bacteria are encountered in an enormous species (micro)diversity in
Yinhu Wang et al.
Biosensors & bioelectronics, 43, 180-185 (2013-01-12)
We report a novel hydrogen peroxide (H2O2) and hydrazine sensor based on low-cost poly(vinylpyrrolidone)-protected silver nanocubes (PVP-AgNCs). The monodisperse silver nanocubes were prepared by adding a trace amount of sodium sulfide in the conventional polyol synthesis for fast reduction of
Tino Wilson Sanchez et al.
Bioorganic & medicinal chemistry, 21(4), 957-963 (2013-01-12)
Human lens epithelium-derived growth factor (LEDGF)/p75 plays an important role in the HIV life cycle by stimulating integrase (IN)-led viral DNA integration into cellular chromosomes. Mechanistic studies show the majority of IN inhibitors chelate magnesium ions in the catalytic active
Michael E Deary et al.
Organic & biomolecular chemistry, 11(2), 309-317 (2012-11-29)
Our recent work has provided new insights into the equilibria and species that exist in aqueous solution at different pHs for the boric acid - hydrogen peroxide system, and the role of these species in oxidation reactions. Most recently, (M.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service