Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900271

Sigma-Aldrich

Manganese(II) sulfate monohydrate

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
99%
grade:
reagent grade
form:
powder
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

powder

pH

3.0-3.5 (20 °C, 50 g/L)

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

target_organs

Brain

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edson R Silva et al.
Experimental parasitology, 125(2), 152-155 (2010-01-30)
In Leishmania, arginase is responsible for the production of ornithine, a precursor of polyamines required for proliferation of the parasite. In this work, the activation kinetics of immobilized arginase enzyme from L. (L.) amazonensis were studied by varying the concentration
Long Peng et al.
Journal of colloid and interface science, 377(1), 13-17 (2012-04-21)
A facile single-source precursor method has been applied for the selective synthesis of MnS nanocrystals (NCs) with well-defined shapes and crystal structures such as hexapod, octahedral, hexagonal shaped α-MnS NCs, and pencil-shaped γ-MnS NCs. The effects of the composition of
Xiaojun Yang et al.
Animal science journal = Nihon chikusan Gakkaiho, 83(6), 474-481 (2012-06-15)
Iron is often found to be of excessive concentrations in laying hens' diets, which may cause antagonistic interactions with other minerals. This study was conducted to investigate how to supplement Zn and Mn in the diets without Fe supplementation. In
Haixing Li et al.
Amino acids, 38(5), 1439-1445 (2009-09-30)
Production of gamma-aminobutyric acid (GABA) was carried out in Erlenmeyer flasks by Lactobacillus brevis NCL912. Traditional methods were first adopted to select the key factors that impact the GABA production to preliminarily determine the suitable concentration ranges of the key
Swatismita Ray et al.
Transgenic research, 21(2), 351-366 (2011-07-05)
Two abiotic stress-inducible upstream regulatory sequences (URSs) from rice have been identified and functionally characterized in rice. NBS-Str1 and BLEC-Str8 genes have been identified, by analysing the transcriptome data of cold, salt and desiccation stress-treated 7-day-old rice (Oryza sativa L.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service