Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900262

Sigma-Aldrich

Thioanisole

Vetec, reagent grade, 98%

Synonym(s):

Methyl phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH3
CAS Number:
Molecular Weight:
124.20
Beilstein/REAXYS Number:
1904179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.587 (lit.)

bp

188 °C (lit.)

mp

−15 °C (lit.)

density

1.057 g/mL at 20 °C (lit.)

SMILES string

CSc1ccccc1

InChI

1S/C7H8S/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

HNKJADCVZUBCPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

156.2 °F - closed cup

flash_point_c

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Yamakita et al.
The journal of physical chemistry. A, 112(47), 12220-12227 (2008-11-05)
Nonresonant Raman spectra and conformational stability are studied for thioanisole (TA) and substituted analogues [4-XTA, X = NO(2) (1), CN (2), H (3), CH(3) (4), and NH(2) (5)] at the 4-position. The ring-substituent (SCH(3)) vibrational modes of out-of-plane bending and
Jeong Sik Lim et al.
Nature chemistry, 2(8), 627-632 (2010-07-24)
Chemical reactions that occur in the ground electronic state are described well by invoking the Born-Oppenheimer approximation, which allows their development to be rationalized by nuclear rearrangements that smoothly traverse an adiabatic potential energy surface. The situation is different, however
Yunho Lee et al.
Inorganic chemistry, 49(19), 8873-8885 (2010-09-09)
To better understand the effect of thioether coordination in copper-O(2) chemistry, the tetradentate N(3)S ligand L(ASM) (2-(methylthio)-N,N-bis((pyridin-2-yl)methyl)benzenamine) and related alkylether ligand L(EOE) (2-ethoxy-N,N-bis((pyridin-2-yl)methyl)ethanamine) have been studied. The corresponding copper(I) complexes, [(L(ASM))Cu(I)](+) (1a) and [(L(EOE))Cu(I)](+) (3a), were studied as were the
Wen Wang et al.
Journal of the American Chemical Society, 131(36), 12892-12893 (2009-08-26)
Magnetic nanoparticles (MNPs) with a core diameter of 30 nm comprising several iron oxide crystals, a poly(glycidyl methacrylate) (PGMA) shell with a thickness of 30 nm, and a surface coated with chloroperoxidase (CPO) were facilely fabricated as a nanobiocatalyst for
Chun Zhu et al.
Physical chemistry chemical physics : PCCP, 14(37), 12800-12806 (2012-08-10)
The electronic and structural features of (oxo)manganese(V) corroles and their catalyzed oxygen atom transfers to thioanisole in different spin states have been investigated by the B3LYP functional calculations. Calculations show that these corrole-based oxidants and their complexes with thioanisole generally

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service