Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900258

Sigma-Aldrich

Diphenyl ether

Vetec, reagent grade, 98%

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
Beilstein/REAXYS Number:
1364620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

>5.86 (25 °C, vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

Vetec

assay

98%

form

solid or liquid

autoignition temp.

1144 °F

expl. lim.

1.5 %

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kostantinos Benekos et al.
Journal of biotechnology, 150(1), 195-201 (2010-07-20)
Plant glutathione transferases (GSTs) superfamily consists of multifunctional enzymes and forms a major part of the plants herbicide detoxification enzyme network. The tau class GST isoenzyme GmGSTU4 from soybean, exhibits catalytic activity towards the diphenyl ether herbicide fluorodifen and is
Ge-Fei Hao et al.
Journal of computer-aided molecular design, 25(3), 213-222 (2011-01-25)
Protoporphyrinogen oxidase (PPO, EC 1.3.3.4), which has been identified as a significant target for a great family of herbicides with diverse chemical structures, is the last common enzyme responsible for the seventh step in the biosynthetic pathway to heme and
Ariana Beste et al.
The Journal of organic chemistry, 74(7), 2837-2841 (2009-03-06)
Lignin is an abundant natural resource that is a potential source of valuable chemicals. Improved understanding of the pyrolysis of lignin occurs through the study of model compounds for which phenethyl phenyl ether (PhCH(2)CH(2)OPh, PPE) is the simplest example representing
Tony Ly et al.
Analytical chemistry, 80(13), 5059-5064 (2008-05-24)
Selective noncovalent adduct protein probing (SNAPP) mass spectrometry was recently developed to study solution-phase conformations of proteins by exploiting the specific affinity between 18-crown-6 ether (18C6) and lysine side chains. To obtain more detailed information about protein tertiary structure, a
Xingyu Wang et al.
ChemSusChem, 5(8), 1455-1466 (2012-05-03)
The conversion of lignin, the most recalcitrant of the biopolymers, is necessary for a carbon-efficient utilization of lignocellulosic materials. In this context, hydrogenolysis of lignin is a process receiving increasing attention. In this report, the solvent effects on the hydrogenolysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service