Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900255

Sigma-Aldrich

Cesium sulfate

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cs2O4S
CAS Number:
Molecular Weight:
361.87
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

form

powder, crystals or chunks

pH

6.0-7.5 (25 °C, 723.7%)

mp

1019 °C (lit.)

density

4.243 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[Cs+].[O-]S([O-])(=O)=O

InChI

1S/2Cs.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

FLJPGEWQYJVDPF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Sheehan et al.
The Biochemical journal, 199(3), 581-589 (1981-12-01)
1. Two proteodermatan sulphate species from bovine sclera (fractions PG-I and PG-II) separable by gel chromatography were studied by isopycnic centrifugations in CsCl and Cs(2)SO(4) both in an analytical and a preparative mode. 2. In CsCl, fraction PG-I formed a
M Bubunenko et al.
Journal of cellular biochemistry, 80(4), 560-570 (2001-02-13)
Two pathways operate during Xenopus oogenesis to localize a small number of RNAs to the vegetal cortex. Correct localization of these RNAs is essential to normal development as the proteins they encode are involved in specifying cell type and in
F M Ashcroft et al.
The Journal of physiology, 359, 269-291 (1985-02-01)
The non-penetrating potentiometric dyes NK2367 and WW375 were used to investigate the effect of inward rectification on the weighted-average tubular membrane potential in single frog muscle fibres voltage clamped using a three-Vaseline-gap method. In 100 mM-K solution, when inward rectification
A K Attri et al.
Analytical biochemistry, 159(1), 88-95 (1986-11-15)
We present a technique for analytical buoyant density sedimentation of nucleic acids which is performed in a preparative ultracentrifuge, in contrast to an analytical ultracentrifuge. Following centrifugation in a preparative rotor, small cylindrical quartz tubes are optically scanned; upon completion
A Miyakawa et al.
Neuroscience, 134(2), 595-604 (2005-06-25)
Effects of neuropeptide Y (NPY) on substantia gelatinosa neurons were investigated in adult rat spinal cord slices using blind whole-cell patch-clamp technique. Bath application of NPY (1 microM) induced a membrane hyperpolarization, resulting in a suppression of the dorsal root

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service