V900253
(±)-1,3-Butanediol
Vetec™, reagent grade, 98%
Synonym(s):
1,3-Butylene glycol
About This Item
grade
reagent grade
vapor density
3.1 (20 °C, vs air)
vapor pressure
0.06 mmHg ( 20 °C)
product line
Vetec™
assay
98%
form
viscous liquid
autoignition temp.
741 °F
expl. lim.
0.34-6.3 % (lit.)
refractive index
n20/D 1.44 (lit.)
pH
6.0-7.0 (20 °C)
bp
203-204 °C (lit.)
mp
-54 °C (lit.)
density
1.005 g/mL at 25 °C (lit.)
SMILES string
CC(O)CCO
InChI
1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3
InChI key
PUPZLCDOIYMWBV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
226.4 °F - closed cup
flash_point_c
108 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service