Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900251

Sigma-Aldrich

Propylene glycol monomethyl ether acetate

Vetec, reagent grade

Synonym(s):

1,2-Propanediol monomethyl ether acetate, 1-Methoxy-2-propyl acetate, MPA, PGMEA, Propylene glycol methyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)CH2OCH3
CAS Number:
Molecular Weight:
132.16
Beilstein/REAXYS Number:
1751656
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor pressure

3.7 mmHg ( 20 °C)

product line

Vetec

autoignition temp.

669 °F

expl. lim.

13.1 %

refractive index

n20/D 1.402 (lit.)

bp

145-146 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

SMILES string

COCC(C)OC(C)=O

InChI

1S/C6H12O3/c1-5(4-8-3)9-6(2)7/h5H,4H2,1-3H3

InChI key

LLHKCFNBLRBOGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles A Conn et al.
Lab on a chip, 14(20), 3968-3977 (2014-08-13)
Foam mobility control and novel oil displacement mechanisms were observed in a microfluidic device representing a porous media system with layered permeability. Foam was pre-generated using a flow-focusing microfluidic device and injected into an oil-wet, oil-saturated 2-D PDMS microfluidic device.
Shaohua Ma et al.
Lab on a chip, 14(18), 3611-3620 (2014-07-30)
The flow topology in moving microdroplets has a significant impact on the behaviour of encapsulated objects and hence on applications of the technology. This study reports on a systematic investigation of the flow field inside droplets moving in a rectangular
Sara Sheikhzadeh et al.
Orphanet journal of rare diseases, 9, 203-203 (2014-12-11)
Echocardiographic upper normal limits of both main pulmonary artery (MPA) diameters (MPA-d) and ratio of MPA to aortic root diameter (MPA-r) are not defined in healthy adults. Accordingly, frequency of MPA dilatation based on echocardiography remains to be assessed in
Simon Braun et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(12), 3641-3646 (2015-03-17)
The dynamics of quantum phase transitions pose one of the most challenging problems in modern many-body physics. Here, we study a prototypical example in a clean and well-controlled ultracold atom setup by observing the emergence of coherence when crossing the
Takuya Miyano et al.
International journal of pharmaceutics, 475(1-2), 504-513 (2014-09-15)
In process analytical technology (PAT) based on near infrared (NIR) spectroscopy, wavenumber selection is crucial to develop an accurate and robust calibration model. The present research proposes new efficient spectral dividing and wavenumber selection methods to significantly reduce the computational

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service